Sitzung fortführen?
Automatische Abmeldung in: 
Sitzung abgelaufen!
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-DUSP8 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
Dual specificity protein phosphatases (DSPs) are implicated in the roles of mitogenic signal transduction regulation as well as cell cycle control.The expression of DUSP1 gene is induced in human skin fibroblasts by oxidative/heat stress as well as growth factors and specifies a protein with structural features similar to members of the non-receptor-type protein-tyrosine phosphatase family. It has significant amino-acid sequence similarity to a Tyr/Ser-protein phosphatase encoded by the late gene H1 of vaccinia virus. Purified DUSP1 protein has intrinsic phosphatase activity, and specifically inactivates mitogen-activated protein (MAP) kinase in vitro by the concomitant dephosphorylation of both its phosphothreonine and phosphotyrosine residues. Additionally, it suppresses the activation of MAP kinase by oncogenic ras in extracts of Xenopus oocytes. Thus, DUSP1 may play an important role in the human cellular response to environmental stress as well as in the negative regulation of cellular proliferation. The Dual specificity protein phosphatase motif is found in 5 isoforms from this gene. DUSP8 encodes a dual specificity MAP kinase phosphatase which has been shown to hydrolyze p-nitrophenylphosphate and inactivate mitogen-activated protein kinases in vitro. The mouse homologue, M3/6, has been demonstrated to be uniquely specific for JNK and p38 kinases. DUSP8 has been mapped to 11p15, a chromosomal region that has been implicated in the development of a number of tumor types, including lung which is a tissue known to express this gene, and loss of heterozygosity has been found in one of eight informative lung tumors studied.

DUSP8 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DUSP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

PRODUKT

Erweitert / Kompakt
Größe VE Katalognummer Menge
50 µg 50 µG PRSI49-273
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol DUSP8
Antigen name Dual specificity phosphatase 8
Reaktivität Human
Konjugation Unconjugated
Klonalität Polyclonal
Wirt Rabbit
Isotyp IgG
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms Dual specificity protein phosphatase 8, HB5, Dual specificity protein phosphatase hVH-5, VH5, HVH-5, HVH8, C11orf81
Immunogen DUSP8 antibody was raised against amino acids 250 - 300 of DUSP8 (Human).
Form Liquid

BEWERTUNG VERFASSEN

Brauchen Sie Hilfe?

  • Was auch immer Sie zu sagen haben, positiv oder negativ, ist uns wichtig.
  • Das Bewerten dauert nur wenige Sekunden
  • Ihre Meinung wird öffentlich zur Verfügung gestellt.

VWR wird Ihren Namen als VWR Nutzer veröffentlichen.
Sie können diesen Namen nutzen oder vorher nochmal ändern und erneut abspeichern.
Wir empfehlen Initialien oder Spitznamen.

Bitte bestätigen Sie die Branche.

Beispiel: Pharma

Bitte bestätigen Sie Ihre Rolle.

Beispiel: Techniker

Bitte bestätigen Sie den Ort

Beispiel: Frankfurt

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

Weitere Angaben

Bitte wählen Sie den gewünschten Artikel

Bitte tragen Sie die Chargennummer ein. Sollte Ihnen diese nicht vorliegen, tragen Sie bitte bei "VWR-Chemikalien" ein "x" ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Kostengünstigere Alternativen prüfen & vergleichen
Wir haben eine kostengünstigere Alternative für Sie gefunden. Ihre Einsparung beträgt  pro Stück.  Sie können bis zu 3 Artikel miteinander vergleichen. Wählen Sie die gewünschten Artikel aus und drücken Sie auf Artikel vergleichen. Zum Ersetzen des ursprünglichen Artkels, geben Sie eine Menge im Alternativprodukt ein und klicken Sie auf Ersetzen.

Ursprünglicher Artikel:

Beschreibung Artikelnummer Verfügbarkeit Einheit Ihr Preis Preis pro Menge