Anti-CCR1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-270EA 480 EUR
PRSI49-270
Anti-CCR1 Rabbit Polyclonal Antibody
Chemokines play important roles in inflammation and critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines. One of the CCR receptors, CCR1, is expressed on neutrophils, monocytes, lymphocytes, and eosinophils and binds the leukocyte chemoattractant and hemopoiesis regulator macrophage-inflammatory protein (MIP-1 ), eotaxin, as well as several other related chemokines. Mice lacking the chemokine receptor CCR1 have defects in neutrophil trafficking and proliferation (5,6). CCR1, a Chemokine Receptor, affects stem cell proliferation and mediates systemic inflammatory responses. This receptor is stimulated by the beta chemokines RANTES, MCP-1, and MIP-1 alpha. Disruption of CCR1 enhances protection from pulmonary inflammation and reduces the development of respiratory distress syndrome in rat.

CCR1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.05% sodium azide, 0.2% gelatin.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR