Anti-BMP10 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-267EA 0 EUR
PRSI49-267
Anti-BMP10 Rabbit Polyclonal Antibody
Bone Morphogenic Protein 10 is a member of the transforming growth factor-beta superfamily. In mouse heart, BMP-10 expression is localized to the trabeculated part of the common ventricular chamber and to the bulbus cordis region, the atrial wall and in the adult right atrium. BMP-10 has recently been implicated in postnatal cardiac hypertrophy.

BMP10 antibody can be used in ELISA starting at 1:10000 - 1:40000, Western Blot starting at 1:500 - 1:3000, and immunohistochemistry starting at 10 - 20 μg/mL.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BMP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR