Sitzung fortführen?
Automatische Abmeldung in: 
Sitzung abgelaufen!
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Anti-BIRC7 Mouse Monoclonal Antibody

Lieferant: ProSci Inc.
The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity. Recently, Kasof and Gomes , Vucic et al. and Ashhab et al. have identified a novel human inhibitor of apoptosis protein (IAP) family member termed Livin or ML-IAP, which contains a single baculoviral IAP repeat (BIR) domain and a COOH-terminal RING finger domain. Livin gene has two splicing variants that contain open reading frames of 298 and 280 amino acids and both contained a single copy of baculovirus IAP repeat (BIR) and RING domain. Both of the isoforms inhibit TNFa induced apoptosis in Jurkat cells. Livin expression is low in adult tissues. It is relatively expressed at a higher level in developmental tissues and in many cancer cells. A novel member in the IAP protein family was recently identified and designated Livin and KIAP for kidney IAP. Livin/XIAP contains a single baculoviral IAP repeat (BIR) domain and a RING finger domain and has two isoforms termed Livin-a and Livin-b. Transfection of Livin in cells resulted in protection from apoptosis induced by FADD, BAX, RIP, RIP3 and DR6. Livin has direct interaction with several caspases including caspase-3, -7, and -9. Livin inhibits the activation of caspase-9 induced by Apaf-1, cytochrome c, and dATP. The two isoforms of Livin appear to have different functions and tissue distributions.

BIRC7 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS containing 0.05% BSA and 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BIRC7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

PRODUKT

Erweitert / Kompakt
Größe VE Katalognummer Menge
50 µg 50 µG PRSI49-265
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen

TECHNISCHE DATEN

Antigen symbol BIRC7
Antigen name Baculoviral IAP Repeat Containing 7
Reaktivität Human
Konjugation Unconjugated
Klonalität Monoclonal
Wirt Mouse
Isotyp IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms LIVIN, Kidney inhibitor of apoptosis protein, Melanoma inhibitor of apoptosis protein, KIAP, RING finger protein 50, RNF50, BIRC7, ML-IAP, Baculoviral IAP repeat-containing protein 7, MLIAP
Immunogen BIRC7 monoclonal antibody was raised against a fusion protein of BIRC7 (Human).
Form Liquid

BEWERTUNG VERFASSEN

Brauchen Sie Hilfe?

  • Was auch immer Sie zu sagen haben, positiv oder negativ, ist uns wichtig.
  • Das Bewerten dauert nur wenige Sekunden
  • Ihre Meinung wird öffentlich zur Verfügung gestellt.

VWR wird Ihren Namen als VWR Nutzer veröffentlichen.
Sie können diesen Namen nutzen oder vorher nochmal ändern und erneut abspeichern.
Wir empfehlen Initialien oder Spitznamen.

Bitte bestätigen Sie die Branche.

Beispiel: Pharma

Bitte bestätigen Sie Ihre Rolle.

Beispiel: Techniker

Bitte bestätigen Sie den Ort

Beispiel: Frankfurt

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

Weitere Angaben

Bitte wählen Sie den gewünschten Artikel

Bitte tragen Sie die Chargennummer ein. Sollte Ihnen diese nicht vorliegen, tragen Sie bitte bei "VWR-Chemikalien" ein "x" ein.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Kostengünstigere Alternativen prüfen & vergleichen
Wir haben eine kostengünstigere Alternative für Sie gefunden. Ihre Einsparung beträgt  pro Stück.  Sie können bis zu 3 Artikel miteinander vergleichen. Wählen Sie die gewünschten Artikel aus und drücken Sie auf Artikel vergleichen. Zum Ersetzen des ursprünglichen Artkels, geben Sie eine Menge im Alternativprodukt ein und klicken Sie auf Ersetzen.

Ursprünglicher Artikel:

Beschreibung Artikelnummer Verfügbarkeit Einheit Ihr Preis Preis pro Menge