Anti-APEX2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-262EA 480 EUR
PRSI49-262
Anti-APEX2 Rabbit Polyclonal Antibody
Antikörper
Apurinic/apyrimidinic (AP) sites occur frequently in DNA by spontaneous hydrolysis, DNA damaging agents or DNA glycosylases that remove specific abnormal bases. ClassII AP endonucleases act to repair this damage by cleaving the phosphodiester backbone. It is primarily located in the nucleus but is also present in mitochondria. This protein may play an important role in both nuclear and mitochondrial base excision repair (BER).

APEX Nuclease antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APEX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Gorilla, Human, Monkey, Pig, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR