Anti-KDM1A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-261EA 514 EUR
PRSI49-261
Anti-KDM1A Rabbit Polyclonal Antibody
LSD1 is a flavin-containing novel nuclear protein having an N-terminal SWIRM domain and a C-terminal FAD-binding motif and an amine oxidase domain. It functions as a transcriptional corepressor and catalyzes the flavin-dependant demethylation of Lys4 of histone3 resulting in the formation of methyl-free lysine and release of formaldehyde. LSD1 has been found to be typically associated with CoREST and histone deacetylases 1 and 4 and participates in the silencing of endogenous neuron-specific genes.

AOF2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KDM1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Macaque, Monkey, Mouse, Rat, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR