Anti-ACVR1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-257EA 480 EUR
PRSI49-257
Anti-ACVR1 Rabbit Polyclonal Antibody
Antikörper
ACVR1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACVR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Xenopus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR