Anti-MCM6 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-255EA 375 EUR
PRSI49-255
Anti-MCM6 Rabbit Polyclonal Antibody
Antikörper
MCM6 antibody can be used in ELISA, Western Blot starting at 1:200, and immunohistochemistry starting at 1:50.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MCM6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Horse, Human, Monkey, Mouse, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR