Anti-LMNA Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-252EA 480 EUR
PRSI49-252
Anti-LMNA Rabbit Polyclonal Antibody
Antikörper
Lamin A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 20 μg/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LMNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR