Anti-LAT Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-251EA 480 EUR
PRSI49-251
Anti-LAT Mouse Monoclonal Antibody
Antikörper
LAT antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 μg/mL, and immunofluorescence starting at 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store LAT antibody at 4 °C.

Type:
Antigen: LAT
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR