Anti-CK19 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-250EA 0 EUR
PRSI49-250
Anti-CK19 Mouse Monoclonal Antibody
KRT19 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Store KRT19 antibody at 4 °C. Store undiluted.

Type:
Antigen: CK19
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR