Anti-GYPA Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-243EA 480 EUR
PRSI49-243
Anti-GYPA Mouse Monoclonal Antibody
Antikörper
Glycophorin A antibody can be used in immunohistochemistry starting at 15 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store Glycophorin A antibody undiluted at 4 °C.

Type:
Antigen: GYPA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR