Anti-FUT4 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-241EA 480 EUR
PRSI49-241
Anti-FUT4 Mouse Monoclonal Antibody
Antikörper
FUT4 catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15). It has been detected in human embryos (5-10 weeks) suggesting a role in development.

FUT4 antibody can be used in immunohistochemistry starting at 5 - 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store FUT4 antibody undiluted at 4 °C.

Type:
Antigen: FUT4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Chimpanzee, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR