Anti-EIF2AK4 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-235EA 480 EUR
PRSI49-235
Anti-EIF2AK4 Rabbit Polyclonal Antibody
Eukaryotic initiation factor-2 alpha (eIF-2 alpha) kinase 4, a Translational Control type protein kinase, is the mammalian homolog of yeast GCN2. Like its yeast counterpart, human GCN2 is believed to inhibit translation by phosphorylating the initiation factor eIF2alpha chain in response to amino acid starvation.

EImmunofluorescence2AK4 antibody can be used in ELISA starting at 1: 10000, Western Blot starting at 1: 500 - 1: 2000, and immunohistochemistry starting at 1:200.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Control
Antigen: EIF2AK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR