Anti-EIF2S1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-234EA 375 EUR
PRSI49-234
Anti-EIF2S1 Rabbit Polyclonal Antibody
Antikörper
EImmunofluorescence2A antibody can be used in ELISA starting at 1: 10000, Western Blot starting at 1: 500 - 1: 2000, immunohistochemistry starting at 1:200, and immunofluorescence starting at 1:200 - 1:1000.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EIF2S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR