Anti-CRP Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-231EA 480 EUR
PRSI49-231
Anti-CRP Rabbit Polyclonal Antibody
CRP antibody can be used in ELISA starting at 1: 10000, Western Blot starting at 1: 500 - 1: 2000, and immunohistochemistry starting at 1:200.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR