Anti-RGS1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-230EA 480 EUR
PRSI49-230
Anti-RGS1 Rabbit Polyclonal Antibody
Regulators of G-protein signaling (RGSs) are a protein family containing a conserved 120 amino acid domain (RGS domain), which bind directly to activated G-alpha subunits (Gai and Gaq) and act as GTPase-activating proteins (GAPs). RGS1 shows a high degree of selectivity to regulate alpha2A-adrenoreceptor-activated G(o1)alpha rather than G(i1)alpha, G(i2)alpha or G(i3)alpha and different capacities to inactivate this G protein.

RGS1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RGS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR