Anti-PRDM5 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-228EA 480 EUR
PRSI49-228
Anti-PRDM5 Rabbit Polyclonal Antibody
Antikörper
PFM2; PR domain containing 5; Official Gene Symbol- PRDM5 Gene ID- 11107(Human) Gene Map Locus- 4q25-q26(Human) PRDM5, a novel member of PR domain protein family, contains a PR-domain at the NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.

PRDM5 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PRDM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR