Anti-NKIRAS1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-224EA 480 EUR
PRSI49-224
Anti-NKIRAS1 Mouse Monoclonal Antibody
NF-κB is silenced in the cytoplasm by an inhibitory protein, IkB. Synthesis of IkBa is autoregulated. IkB proteins are phosphorylated by IkB kinase complex consisting of at least three proteins, IKK1/a, IKK2/b, and IKK3/g (3-6). External stimuli such as tumor necrosis factor or other cytokines results in phosphorylation and degradation of IkB releasing NF-κB dimers. NF-κB dimer subsequently translocates to the nucleus and activates target genes. Six members of IkB family members have been identified. One of the first genes induced following NF-κB activation is IkBa. Recently, two proteins, kB-Ras1 and kB-Ras2 have been identified. These two proteins interact with PEST domains of IkBa and IkBb and decrease their rate of degradation.

NKIRAS1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NKIRAS1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Dog, Gorilla, Goat, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR