Anti-NFATC1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-223EA 480 EUR
PRSI49-223
Anti-NFATC1 Rabbit Polyclonal Antibody
Antikörper
NFAT (nuclear factor of activated T cell transcription factors play critical roles in gene transcription during immune responses. The NFAT complex is made up of NFATc (NFATc1-4) protein and activator protein-1 (AP-1) transcription factor. NFAT proteins are regulated by Ca2+. NFATc1 is a substrate protein phosphatase calcineurin, a critical mediator of calcium signals during T-cell activation. NFATc1 activation requires the translocation of the transcription factor into the nucleus, a process that is mediated by calcineurin.

NFATC1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NFATC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR