Anti-CDK7 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-217EA 375 EUR
PRSI49-217
Anti-CDK7 Rabbit Polyclonal Antibody
Antikörper
CDK7, a CDC2/CDK type protein kinase, is the catalytic component of the Cdk-activating kinase (CAK) which acts as a regulator of cell cycle progression. CDK7 complexes with cyclin H and MAT1 to form CAK, and this multi-subunit protein phosphorylates the cyclin-dependent protein kinases CDC2/CDK1, CDK2, CDK4 and CDK6. CAK also associates with the transcription factor IIH (TFIIH) which functions in transcription initiation and DNA repair. TFIIH has been shown to be regulated by CDK8/cyclin C.

CDK7 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 5 μg/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDK7
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR