Anti-CD99 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-211EA 375 EUR
PRSI49-211
Anti-CD99 Mouse Monoclonal Antibody
Antikörper
CD99 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 2.5 μg/mL, immunofluorescence, and immunoprecipitation.

Affinity Chromatography

Store CD99 antibody undiluted at 4 °C. Do not freeze.

Store CD99 antibody undiluted at 4 °C. Do not freeze.

Type:
Antigen: CD99
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR