Anti-CD74 Mouse Monoclonal Antibody (Biotin)

Lieferant: ProSci
PRSI49-209EA 375 EUR
PRSI49-209
Anti-CD74 Mouse Monoclonal Antibody (Biotin)
Antikörper
CD74 antibody can be used in ELISA starting at 1:5000 - 1:40000, Western Blot starting at 1:5000 - 1:40000, and immunohistochemistry starting at 5 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store CD74 antibody undiluted at 4 °C.

Type:
Antigen: CD74
Clonality: Monoclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR