Anti-CD63 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-207EA 375 EUR
PRSI49-207
Anti-CD63 Mouse Monoclonal Antibody
Antikörper
CD63 antibody can be used in immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store CD63 antibody undiluted at 4 °C.

Type:
Antigen: CD63
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR