Anti-CD40 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-201EA 480 EUR
PRSI49-201
Anti-CD40 Mouse Monoclonal Antibody
Antikörper
CD40 antibody can be used in immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

PBS, pH 7.2, 0.09% sodium azide.

Store CD40 antibody undiluted at 4 °C.

Type:
Antigen: CD40
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR