Anti-BSG Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-195EA 480 EUR
PRSI49-195
Anti-BSG Mouse Monoclonal Antibody
Antikörper
Basigin antibody can be used in immunohistochemistry starting at 10 μg/mL, and immunohistochemistry in frozen tissues starting at 1:400 - 1:600.

Affinity Chromatography

PBS, pH 7.2, 0.09% sodium azide.

Store Basigin antibody undiluted at 4 °C.

Type:
Antigen: BSG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR