Anti-MMP1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-169EA 530 EUR
PRSI49-169
Anti-MMP1 Rabbit Polyclonal Antibody
Antikörper
MMP1 is a Metallopeptidase M10A type protease. MMP1, also known as collagenase, is a secreted protein. Collagenase is the only enzyme able to initiate breakdown of the interstitial collagens, types I, II, and III. The fact that the collagens are the most abundant proteins in the body means that collagenase plays a key role in the remodeling that occurs constantly in both normal and diseased conditions.

MMP1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, pH 7.4, 0.05% sodium azide.

PBS, pH 7.4, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: MMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR