Anti-VCP Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-154EA 480 EUR
PRSI49-154
Anti-VCP Mouse Monoclonal Antibody
Antikörper
VCP antibody can be used in immunohistochemistry starting at 20 μg/mL, and immunofluorescence.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: VCP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR