Anti-TNFSF13B Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-150EA 480 EUR
PRSI49-150
Anti-TNFSF13B Mouse Monoclonal Antibody
Antikörper
Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated BAFF (for B cell Activating Factor belonging to the TNF Family), BLyS (for B Lymphocyte Stimulator), TALL-1 (for TNF- and ApoL-related Leukocyte-expressed Ligand), and THANK (for TNF Homologue that Activate Apoptosis, NF-κB and c-jun N-terminal Kinase). BAFF/BLyS was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF and its receptors are essential for B cell development, survival, and humoral immune responses. BAFF is involved in the development of autoimmune diseases including systemic lupus erythaematosus and rheumatoid arthritis.

TNFSF13B antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store TNFSF13B antibody at 4 °C. Do not freeze.

Type:
Antigen: TNFSF13B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR