Anti-TAF1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-148EA 375 EUR
PRSI49-148
Anti-TAF1 Rabbit Polyclonal Antibody
Antikörper
TAF1 antibody can be used in Western Blot, immunohistochemistry starting at 5 μg/mL, immunocytochemistry, and immunoprecipitation

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Bovine, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR