Anti-STMN1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-146EA 0 EUR
PRSI49-146
Anti-STMN1 Rabbit Polyclonal Antibody
Stathmin 1 antibody can be used in ELISA starting at 1:1000 - 1:5000, Western Blot, immunohistochemistry starting at 10 μg/mL, and immunoprecipitation

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR