Anti-STAT4 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI49-145EA 480 EUR
PRSI49-145
Anti-STAT4 Rabbit Polyclonal Antibody
Antikörper
STAT4 is a transcription factor involved in promoting cell-mediated immunity. It is a key molecule in the intracellular signal transduction cascade activated in response to IL-12 stimulation. Following phosphorylation in response to IL-12 or IFN-alpha STAT4 dimerizes, translocates to the nucleus, and activates gene expression.

STAT4 antibody can be used in immunohistochemistry starting at 10 μg/mL, immunoprecipitation, and flow cytometry starting at 1:50 - 1:100.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR