Anti-PCNA Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-126EA 480 EUR
PRSI49-126
Anti-PCNA Mouse Monoclonal Antibody
Antikörper
PCNA antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store PCNA antibody at 4 °C. Do not freeze.

Type:
Antigen: PCNA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR