Anti-NCAM1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-121EA 480 EUR
PRSI49-121
Anti-NCAM1 Mouse Monoclonal Antibody
Antikörper
NCAM1 antibody can be used in immunohistochemistry starting at 5 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store NCAM1 antibody at 4 °C.

Type:
Antigen: NCAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR