Anti-LCP2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-118EA 480 EUR
PRSI49-118
Anti-LCP2 Mouse Monoclonal Antibody
LCP2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LCP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR