Anti-MAP2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-111EA 480 EUR
PRSI49-111
Anti-MAP2 Mouse Monoclonal Antibody
Antikörper
MAP2 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MAP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR