Anti-VIM Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-109EA 480 EUR
PRSI49-109
Anti-VIM Mouse Monoclonal Antibody
Antikörper
Vimentin antibody can be used in ELISA, and immunohistochemistry starting at 15 - 20 μg/mL.

Size Exclusion Chromatography

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: VIM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Chicken, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR