Anti-TUBA1B Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-107EA 480 EUR
PRSI49-107
Anti-TUBA1B Mouse Monoclonal Antibody
Antikörper
Precipitation

Precipitation

PBS, 15 mM sodium azide, approx., pH 7.4.

Store TUBA antibody at 4 °C. Do not freeze.

Type:
Antigen: TUBA1B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR