Anti-TUBB Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-106EA 480 EUR
PRSI49-106
Anti-TUBB Mouse Monoclonal Antibody
Antikörper
TUBB antibody can be used in Western Blot starting at 1 - 2 μg/mL, immunohistochemistry starting at 1 - 2 μg/mL, immunoprecipitation starting at 2 µg/mg lysate, and immunofluorescence.

Precipitation

Tris buffered saline (TBS), 15 mM sodium azide, approx., pH 8.0.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: TUBB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR