Anti-BSG Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-103EA 480 EUR
PRSI49-103
Anti-BSG Mouse Monoclonal Antibody
Antikörper
Basigin antibody can be used in ELISA, Western Blot starting at 1 - 2 μg/mL, immunohistochemistry starting at 5 μg/mL, immunofluorescence, and immunoprecipitation.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BSG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR