Anti-CD44 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-101EA 375 EUR
PRSI49-101
Anti-CD44 Mouse Monoclonal Antibody
Antikörper
CD44 antibody can be used in ELISA, Western Blot starting at 1 - 2 μg/mL, immunohistochemistry starting at 10 μg/mL, immunocytochemistry, and immunofluorescence.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD44
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR