Anti-BSG Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-100EA 480 EUR
PRSI49-100
Anti-BSG Mouse Monoclonal Antibody
Antikörper
Basigin antibody can be used in immunohistochemistry starting at 5 μg/mL.

Protein A Column

PBS, 15 mM sodium azide, pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BSG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR