Anti-SNAP25 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-099EA 480 EUR
PRSI49-099
Anti-SNAP25 Mouse Monoclonal Antibody
Antikörper
SNAP25 antibody can be used in Western Blot, and immunohistochemistry starting at 5 μg/mL.

Supernatant

PBS, pH 7.2, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SNAP25
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Human, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR