Anti-CYCS Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-095EA 480 EUR
PRSI49-095
Anti-CYCS Mouse Monoclonal Antibody
Antikörper
Cytochrome c antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.2, 50% glycerol, 0.09% azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CYCS
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR