Anti-CASP7 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-092EA 514 EUR
PRSI49-092
Anti-CASP7 Mouse Monoclonal Antibody
Caspase 7 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS (10 mM sodium phosphate, pH 7.2, 50 mM NaCl, 0.1 mM PMSF)

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CASP7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR