Anti-DIABLO Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-090EA 480 EUR
PRSI49-090
Anti-DIABLO Mouse Monoclonal Antibody
SMAC antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, pH 7.2, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DIABLO
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR