Anti-ALPL Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-088EA 480 EUR
PRSI49-088
Anti-ALPL Mouse Monoclonal Antibody
Antikörper
ALPL antibody can be used in Western Blot, and immunohistochemistry starting at 1 - 2 μg/mL.

Protein A Column

PBS, pH 7.2, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ALPL
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR