Anti-AIF1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-087EA 480 EUR
PRSI49-087
Anti-AIF1 Mouse Monoclonal Antibody
IBA1 antibody can be used in immunohistochemistry starting at 1:50, and immunofluorescence.

Affinity Purified

PBS pH 7.2, 10mg/mL BSA, 0.09% sodium azide

Store IBA1 antibody at 4 °C.

Type:
Antigen: AIF1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR