Anti-Troponin T type 3 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-084EA 480 EUR
PRSI49-084
Anti-Troponin T type 3 Mouse Monoclonal Antibody
Antikörper
TNNT3 antibody can be used in Western Blot starting at 5 - 10 μg/mL, immunohistochemistry starting at 5 μg/mL, immunoprecipitation starting at 10 µg/mg lysate, and immunofluorescence.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Troponin T type 3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Quail
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR