Anti-MYL2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI49-073EA 480 EUR
PRSI49-073
Anti-MYL2 Mouse Monoclonal Antibody
Antikörper
MYL2 antibody can be used in ELISA starting at 1:312500, Western Blot starting at 2.5 μg/mL, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MYL2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR